Learn More
Electron Microscopy Sciences High Heat Cautery Set
SDP
Catalog No. 5029830
Encompass_Preferred
The cautery comes with fully changeable tungsten carbide tips and batteries. Dual power: 1 AA battery for lower power and 2 AA batteries for higher power. Fully autoclavable. Stainless steel construction. Generates up to 1000 deg C, set includes cautery handle and tips.
Catalog No. 50-298-30 Supplier Electron Microscopy Sciences Supplier No. 7297631
The cautery comes with fully changeable tungsten carbide tips and batteries. Dual power: 1 AA battery for lower power and 2 AA batteries for higher power. Fully autoclavable. Stainless steel construction. Generates up to 1000 deg C, set includes cautery handle and tips.