Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Beverage Analysis Standards, High Level, 250 mg/L Caffeine, 100 mL

Catalog No. 502392843 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

These High Level Beverage Analysis Standards provide a 100 mL solution of caffeine (250 mg/L) and acesulfame-K (250 mg/L). Designed for high-caffeine beverages like energy drinks, they are crucial for LC-MS system calibration to increase quantitation precision and accuracy, ensuring proper equipment function for precise analysis before experiments.

  • Separation mode: Reversed phase
  • Detection method: UV
  • Liquid formulation for room temperature storage and ambient shipping
  • Designed for LC systems and quantitative analysis
  • Shelf life of 12 months
  • Volume: 100 mL
  • Type: Calibration standards
  • Application: System calibration for beverages with high caffeine content
  • Increases precision and accuracy of quantitation
  • Supports detection before experiments
  • Useful for cleaning and benchmarking LC-MS equipment performance

Catalog No. 50-239-2843 Supplier Waters Corp Supplier No. 186007232
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

These High Level Beverage Analysis Standards provide a 100 mL solution of caffeine (250 mg/L) and acesulfame-K (250 mg/L). Designed for high-caffeine beverages like energy drinks, they are crucial for LC-MS system calibration to increase quantitation precision and accuracy, ensuring proper equipment function for precise analysis before experiments.

  • Separation mode: Reversed phase
  • Detection method: UV
  • Liquid formulation for room temperature storage and ambient shipping
  • Designed for LC systems and quantitative analysis
  • Shelf life of 12 months
  • Volume: 100 mL
  • Type: Calibration standards
  • Application: System calibration for beverages with high caffeine content
  • Increases precision and accuracy of quantitation
  • Supports detection before experiments
  • Useful for cleaning and benchmarking LC-MS equipment performance

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.