Learn More
Carbosynth LLC. High Load Streptavidin Poly-HRP Conjugate for use in immunoassays
SDP
Catalog No. 501253506
Encompass_Preferred
Streptavidin Poly-HRP Conjugate is streptavidin biotin-binding protein that is conjugated with polymers of horseradish peroxidase, enabling signal amplification and detection of biotinylated antibodies.
Catalog No. 50-125-3506 Supplier Carbosynth LLC. Supplier No. 85R2021MG
Streptavidin Poly-HRP Conjugate is streptavidin biotin-binding protein that is conjugated with polymers of horseradish peroxidase, enabling signal amplification and detection of biotinylated antibodies.