Learn More
Mopec High/Low Audio/Visual Alarm
SDP
Catalog No. 22000159 Shop All Mopec Products
Change view
Click to view available options
Temperature Range (Metric):
-50°C to +70°C
Catalog No. Temperature Range (Metric)
22-000-159 -50°C to +70°C
1 options
Catalog No. 22-000-159 Supplier Mopec Supplier No. KO001
Add to Cart
Add to Cart
This solution provides temperature alarms, door ajar alarms, AC failure alarms and panic alarms.
  • Compact design for flush or surface mounting in a single gang weatherproof enclosure
  • Simple 3 button user interface
  • High and low alarm temperature setpoints
  • Readout in °F or °C with automatic scale conversion
  • Adjustable alarm delays
Specifications
Temperature Range (Metric) -50°C to +70°C
Type Temperature Sensor