Learn More
Waters Corp Waters Corporation UPLC Columns Kit, High and Low pH
Catalog No. 50791058 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The UPLC Columns in these Method Development Kits offer a broad range of selectivity to facilitate various method-development approaches, aiding in finding a reliable column for desired separation results. This specific kit is designed to maximize separation selectivity by exploring both low and high mobile-phase pH conditions.
  • Designed for high and low mobile-phase pH exploration
  • Covers a broad range of selectivity
  • Column dimensions: 3.0 x 50 mm
  • Particle size: 1.7 μm
  • Includes BEH C18, BEH C8, BEH Shield RP18, BEH Phenyl stationary phases
  • Supplied as a pack of 4 columns
Catalog No. 50-791-058 Supplier Waters Corp Supplier No. 176001881
Add to Cart
Add to Cart
Request a Quote
The UPLC Columns in these Method Development Kits offer a broad range of selectivity to facilitate various method-development approaches, aiding in finding a reliable column for desired separation results. This specific kit is designed to maximize separation selectivity by exploring both low and high mobile-phase pH conditions.
  • Designed for high and low mobile-phase pH exploration
  • Covers a broad range of selectivity
  • Column dimensions: 3.0 x 50 mm
  • Particle size: 1.7 μm
  • Includes BEH C18, BEH C8, BEH Shield RP18, BEH Phenyl stationary phases
  • Supplied as a pack of 4 columns