Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streamlight™ Enduro™ Pro Haz-Lo™ Headlamp

Catalog No. 19028337 Shop All Streamlight Products
Change view
Click to view available options
Color:
Yellow
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-028-337 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-028-337 Supplier Streamlight™ Supplier No. 45680
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Low-profile industrial headlamp with Class I, Division 1 safety ratings for use in hazardous locations. Three lighting modes cover a variety of tasks: spot for a focused beam, flood for soft area illumination that won't tire your eyes, and a spot/flood combo for even more light output.

  • Tough thermoplastic construction with elastomer over mold
  • Unbreakable polycarbonate lenses
  • Available in Safety Yellow
  • Includes rubber strap and 3M Dual Lock™ for strapless mounting to hard hats
  • Push button switch; Click switch to step through each mode, Spot, Flood, Spot/Flood Combo, and OFF
  • IPX7 rated design; waterproof to 1 meter for 30 minutes
  • 2m impact resistance tested
  • 45 degree tilting head, especially handy for reading

Specifications

Light Source 2 x White LED
Weight (English) 3.5 oz.
Weight (Metric) 0.099 kg
Battery Type Three AAA alkaline batteries (inlcuded). Mechanical battery polarity protected.
Beam Distance 73 m
Certifications/Compliance Class I, II, Division 1, Groups A, B, C, D, E, F, G; Class III; Class 1, Zone 0, Group IIC, Exia; T-Code: T4 (approved for use with Energizer E92, Duracell MN2400, Rayovac NO.824, and Panasonic LR03PA).MSHA Permissible Flashlight, Approval No. 20-A190001-0. Tested for Intrinsic Safety in Methane-Air mixtures only.Meets applicable European Community directives.
Color Yellow
Dimensions (L x W x H) 3.8 x 5.68 x 5.1 cm (1.5 x 2.19 x 2 in.)
Includes AAA alk. batteries, elastic headstrap, rubber hard hat strap, and 3M Dual Lock fasteners
Material Thermoplastic
Output
Spot and flood beams:
Spot: 160 lumens; 1,320 candela; 73m beam; runs 5 hours 
Flood: 160 lumens; 100 candela; 20m beam; runs 5 hours
Spot/flood combo: 235 lumens; runs 4.75 hours
Product Type Headlamp
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.