Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streamlight™ High Lumen Rechargeable Portable Scene Light II

Catalog No. 19028338 Shop All Streamlight Products
Change view
Click to view available options
Color:
Red
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-028-338 Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-028-338 Supplier Streamlight™ Supplier No. 46000
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bright waterproof area light requires zero depolyment time and provides 10,000 lumens. With a rotating head to aim the light where you need it, this cordless scene light eliminates tripping hazards common with corded lighting.Plus, portability from job to job is a snap.

  • Turn to a higher power with the rechargeable Portable Scene Light II
  • This super bright waterproof area light requires zero depolyment time and provides 10,000 lumens to light your way
  • Ideal for field maintenance, construction sites, first responders, military operations, and general scene lighting. Stack up to three units for an intense light tower
  • With a rotating head to aim the light where you need it, this cordless scene light eliminates tripping hazards common with corded lighting options
  • Plus, portability from job to job is a snap
  • 10,000 lumens on high
  • Stable stacking of 3 units for high intensity light tower
  • Rotating head allows for precise aim of beam
  • Five levels of light output to achieve desired runtime
  • IPX7 waterproof to 1 meter for 30 min
  • Remaining runtime is displayed
  • 11 hours of runtime at 20% light intensity

Specifications

Voltage 10.89 V
Weight (English) 15.38 lb.
Weight (Metric) 6.10 kg
Battery Run Time 2 Hrs (High), 11 Hrs (Low)
Battery Type Lithium-ion
Certifications/Compliance IPX 7 tested CEC CEC Compliant
Color Red
Includes Battery, 120 V AC & 12 V DC charging cords
Length (English) 16 in.
Lumen 10000 lm (High), 2300 lm (Low)
Material Thermoplastic
Warranty 2 year warranty with proof of purchase
Product Type Headlamp
Show More Show Less

IPX 7 tested CEC CEC Compliant

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.