Learn More
Perkin Elmer US LLC High Matrix Quartz Torch for NexION 1000/2000
Catalog No. 502104155
Encompass_Preferred
This torch provides a longer lifetime when analyzing high matrix samples.
Catalog No. 50-210-4155 Supplier Perkin Elmer US LLC Supplier No. N8145450
This torch provides a longer lifetime when analyzing high matrix samples.