Learn More
Mercodia Inc Diabetes Antigen Control, Rat and Mouse, Low Medium High
SDP
Catalog No. NC1867959
Encompass_Preferred
Control set containing rat and mouse insulin and rat C-peptide to be used in the rat and mouse insulin kits as well as in Rat C-peptide ELISA.
Catalog No. NC1867959 Supplier Mercodia Inc Supplier No. 10122001
Control set containing rat and mouse insulin and rat C-peptide to be used in the rat and mouse insulin kits as well as in Rat C-peptide ELISA.