Learn More
Gelest Inc GELEST hPDMS: High Modulus Reprographic Silicone
Supplier:  Gelest Inc PP2RG071KG
Encompass
This item has a minimum qty of 2 per supplier requirements.
hPDMS is a clear molding and encapsulation PDMS with a higher modulus than conventional silicones. This 'hard' PDMS addresses the issues of compliant PDMS in soft lithography applications.
Catalog No. NC1197697