Learn More
Cole-Parmer Digi-Sense Oven Verification Thermometer; 50 to 200C, PFA Coated
Catalog No. NC0274095 Shop All Cole-Parmer Products
Encompass_Preferred
Accurately monitor temperatures in freezers, refrigerators, incubators, ovens, and other environments where stability is required. These thermometers are supplied in a media-filled unbreakable plastic bottle and can be removed from the bottle for recalibration. Use the magnetic attachment with pressure-sensitive backing to attach the bottle to a metal wall or door. Thermometers feature permanently fused markings, blue spirit liquid, white background glass; are also individually serial numbered. Include an statement of accuracy and a triangular-style antiroll fitting.
Catalog No. NC0274095 Supplier Cole-Parmer Supplier No. 9030073
Accurately monitor temperatures in freezers, refrigerators, incubators, ovens, and other environments where stability is required. These thermometers are supplied in a media-filled unbreakable plastic bottle and can be removed from the bottle for recalibration. Use the magnetic attachment with pressure-sensitive backing to attach the bottle to a metal wall or door. Thermometers feature permanently fused markings, blue spirit liquid, white background glass; are also individually serial numbered. Include an statement of accuracy and a triangular-style antiroll fitting.