Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex I/P Precision Pump Tubing, C-Flex, I/P 82; 100 ft
Catalog No. NC0256909
Encompass
Masterflex C-Flex Precision pump tubing offers enhances biocompatibility with longer pumping life. This tubing is nonpyrogenic, noncytotoxic, and nonhemolytic ¿ protecting the purity of your fluid. Heat sealable and weldable, C-Flex tubing is ideal for biopharmaceutical processing and storage and botanical production. Spools contain continuous pump tubing, allowing you to cut the exact length needed and eliminating waste.
Catalog No. NC0256909 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0642782
Masterflex C-Flex Precision pump tubing offers enhances biocompatibility with longer pumping life. This tubing is nonpyrogenic, noncytotoxic, and nonhemolytic ¿ protecting the purity of your fluid. Heat sealable and weldable, C-Flex tubing is ideal for biopharmaceutical processing and storage and botanical production. Spools contain continuous pump tubing, allowing you to cut the exact length needed and eliminating waste.