Learn More
Sheldon Manufacturing Inc HIGH PERFORMANCE ANAEROBIC CHA
SDP
Catalog No. NC2746867
Encompass_Preferred
HIGH PERFORMANCE ANAEROBIC CHAMBER 400 PETRI PLATE CAPACITY INCUBATOR, 115V; Hepa filtered work chamber, flip-up front flange,multi-funtion large touch screen control, single plate entry system.
Catalog No. NC2746867 Supplier Sheldon Manufacturing Inc Supplier No. BAA400HP23
Add to Cart
Add to Cart
Request a Quote
HIGH PERFORMANCE ANAEROBIC CHAMBER 400 PETRI PLATE CAPACITY INCUBATOR, 115V; Hepa filtered work chamber, flip-up front flange,multi-funtion large touch screen control, single plate entry system.