Learn More
Cole-Parmer Speirs Robertson AMD-SR-120X75-SS AMD-SR Instrument Table, Sorbothane Isolators; 120x75 cm
Catalog No. NC1505173 Shop All Cole-Parmer Products
Encompass
Save time and money by making sure your expensive high performance instruments are not underperforming due to a poor working environment. Vibrations limit the performance of sensitive equipment and manufacturer’s specifications usually state that performance depends on working in the right environment. The tables remove these vibrations allowing you to get the performance you expect. Active tables require a supply of clean dry compressed air of at least 5Bar (10 Bar max) via a 6mm OD airline. Platforms are simple to install, easy to use and maintenance free.
Catalog No. NC1505173 Supplier Cole-Parmer Supplier No. 1150035
Save time and money by making sure your expensive high performance instruments are not underperforming due to a poor working environment. Vibrations limit the performance of sensitive equipment and manufacturer’s specifications usually state that performance depends on working in the right environment. The tables remove these vibrations allowing you to get the performance you expect. Active tables require a supply of clean dry compressed air of at least 5Bar (10 Bar max) via a 6mm OD airline. Platforms are simple to install, easy to use and maintenance free.