Learn More
BUCHI High Performance Condenser Glass Assembly
SDP
Catalog No. 05405080 Shop All BUCHI Products
Change view
Click to view available options
For Use With (Equipment):
R-300, R-210, R-215
Catalog No. For Use With (Equipment)
05-405-080 R-300, R-210, R-215
1 options
Catalog No. 05-405-080 Supplier BUCHI Supplier No. 11066562
Add to Cart
Add to Cart
Request a Quote
Includes a 1 liter receiving flask, the required tubing, and a ball joint clamp. For use with Rotavapor R-300, features a P+G safety coating.
Recommended in the Following Cases:
  • For reduction of solvent emissions
  • For operating at higher temperature difference (ΔT>20°C) and lower pressure values
  • For distilling low boiling solvents
  • If faster process and greater distillation rate are required
Specifications
Material Borosilicate Glass (3.3)
For Use With (Equipment) R-300, R-210, R-215
Includes Receiving Flask (1 L), Required Tubing, and a Ball Joint Clamp
Material (Coating) Plastic, Glass