Learn More
Fritsch Milling And Sizing Inc PULVERISETTE 14 Pl, High-Performance Cyclone Separator Made Of Stainless Steel 304 for Sample Exhaustion, Incl. Sample Glass 1000 Ml
SDP
Catalog No. 501533999
Encompass
The compact FRITSCH high-performance Cyclone separator for PULVERISETTE 14 premium line, which is made completely out of stainless steel 304, is particularly indispensable in the analytical sector and the food and pharmaceutical industries. Due to its high surface quality, it offers enhanced resistance to corrosive media such as alkalis and acids, but particularly to media containing chlorides – and is especially easy to clean with a wide range of possible cleaning agents, without leaving any residues. The Cyclone separator can also be fully dismantled, meaning that it can be completely emptied, flooded and sterilised. Your advantage: reliable protection against cross-contamination.
Catalog No. 50-153-3999 Supplier Fritsch Milling And Sizing Inc Supplier No. 14.4807.00
Add to Cart
Add to Cart
Request a Quote
The compact FRITSCH high-performance Cyclone separator for PULVERISETTE 14 premium line, which is made completely out of stainless steel 304, is particularly indispensable in the analytical sector and the food and pharmaceutical industries. Due to its high surface quality, it offers enhanced resistance to corrosive media such as alkalis and acids, but particularly to media containing chlorides – and is especially easy to clean with a wide range of possible cleaning agents, without leaving any residues. The Cyclone separator can also be fully dismantled, meaning that it can be completely emptied, flooded and sterilised. Your advantage: reliable protection against cross-contamination.