Learn More
Fritsch Milling And Sizing Inc High-Performance Cyclone Separator Made Of Stainless Steel 304 for Sample Exhaustion, Incl. Sample Glass 1000 Ml, P-19
SDP
Catalog No. 501534051
Encompass
The sample exhaust with the high performance Cyclone separator made of stainless steel 304 for PULVERISETTE 19 ensures with its powerful vacuum function, based on negative pressure a simple feeding, fast throughput and good cooling and is especially recommended for analytical requirements,  the food and pharmaceutical industry, as well as absolutely necessary in combination with the corrosion-resistant stainless steel version of the Universal Cutting Mill PULVERISETTE 19 (order no. 19.2040.00)
Catalog No. 50-153-4051 Supplier Fritsch Milling And Sizing Inc Supplier No. 19.1907.00
Add to Cart
Add to Cart
Request a Quote
The sample exhaust with the high performance Cyclone separator made of stainless steel 304 for PULVERISETTE 19 ensures with its powerful vacuum function, based on negative pressure a simple feeding, fast throughput and good cooling and is especially recommended for analytical requirements,  the food and pharmaceutical industry, as well as absolutely necessary in combination with the corrosion-resistant stainless steel version of the Universal Cutting Mill PULVERISETTE 19 (order no. 19.2040.00)