Learn More
DWK Life Sciences Wheaton™ Crimping Jaw Set for High Performance Crimping Tool
SDP
Catalog No. 02915019 Shop All DWK Life Sciences Products
Change view
Click to view available options
Size:
8 mm
11 mm
13 mm
20 mm
For Use With (Application):
Crimping
Crimping flip-off caps
Decapping
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size For Use With (Application)
02-915-019 13 mm Crimping
02-915-014 11 mm Crimping
02-915-015 11 mm Decapping
02-915-016 20 mm Crimping
02-915-017 20 mm Decapping
02-915-018 8 mm Crimping
02-915-020 13 mm Decapping
02-915-021 13 mm Crimping flip-off caps
02-915-022 20 mm Crimping flip-off caps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 02-915-019 Supplier DWK Life Sciences Supplier No. W225761
Only null left
Add to Cart
Add to Cart
Interchangeable jaws allow users to choose a jaw that best fits their needs.
Available as crimper and decappers which fit standard lab vials seals with aluminum or two-part aluminum/steel caps
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) High performance crimping tool
Size 13 mm
For Use With (Application) Crimping