Learn More
Perkin Elmer US LLC High powered Crimper tool (without jaws)
Catalog No. 502111803
Encompass_Preferred
Features: Powerful motor with a fast cycle Long lasting Lithium Ion battery Capable of supporting 20 mm and 11 mm Jaw Sets Enough power for steel and metal caps Able to store multiple programs for different types of caps and seals Advanced charging system with warning light Can be operated while charging Convenient, easy to use controls Optional base and mounting kit Includes: 12V power supply Strip-end power cord Crimper DOC Quick Start Guide CD with multilingual instructions
Catalog No. 50-211-1803 Supplier Perkin Elmer US LLC Supplier No. N9304510
Add to Cart
Add to Cart
Request a Quote
Features: Powerful motor with a fast cycle Long lasting Lithium Ion battery Capable of supporting 20 mm and 11 mm Jaw Sets Enough power for steel and metal caps Able to store multiple programs for different types of caps and seals Advanced charging system with warning light Can be operated while charging Convenient, easy to use controls Optional base and mounting kit Includes: 12V power supply Strip-end power cord Crimper DOC Quick Start Guide CD with multilingual instructions