Learn More
Azzota Corp High Precision Balances 120-32
Catalog No. 5000887249
Encompass_Preferred
Certified high precision analytical balances Accuracy 0 1mg Range 120 to 320 g Stainless steel tray 80 mm
Catalog No. 50-008-87249 Supplier Azzota Corp Supplier No. AUM310I
Certified high precision analytical balances Accuracy 0 1mg Range 120 to 320 g Stainless steel tray 80 mm