Learn More
PCE AMERICAS INC Analytical moisture balance scale with a weighing capacity of up to 60 g with a readability of 0.0001 g.
SDP
Catalog No. NC3654896
Encompass_Preferred
"The high precision moisture analyzer balance enables moisture determinations with a resolution of 0.001 %. The operation of this moisture analyzer balance is very simple. Set the drying temperature, place the sample and start drying.Weighing range: 60 gReadability: 0.1 mg = 0.0001 g / 0.001%.Drying temperature up to 160°CLive graph of moisture contentMoisture content %Dry content %ATRO %WeightProduct database for drying parametersRS232 / USB interfaceStorage on USB stick possible"
Catalog No. NC3654896 Supplier PCE AMERICAS INC Supplier No. PCEMA 60XT
"The high precision moisture analyzer balance enables moisture determinations with a resolution of 0.001 %. The operation of this moisture analyzer balance is very simple. Set the drying temperature, place the sample and start drying.Weighing range: 60 gReadability: 0.1 mg = 0.0001 g / 0.001%.Drying temperature up to 160°CLive graph of moisture contentMoisture content %Dry content %ATRO %WeightProduct database for drying parametersRS232 / USB interfaceStorage on USB stick possible"