Learn More
Qosina Corp HIGH PRES.TUBING LINE,LUER,30"
Catalog No. NC1449571
Encompass
This item has a minimum qty of 5 per supplier requirements.
High Pressure Tubing Line, Female Luer Lock, Male Luer Lock0.071 inch ID x 0.188 inch OD (1.8 mm x 4.8 mm); 30 inch Length (762 mm); 1200 psi
Catalog No. NC1449571 Supplier Qosina Corp Supplier No. 33012
This item has a minimum qty of 5 per supplier requirements.
High Pressure Tubing Line, Female Luer Lock, Male Luer Lock0.071 inch ID x 0.188 inch OD (1.8 mm x 4.8 mm); 30 inch Length (762 mm); 1200 psi