Learn More
Beckman Coulter MET ONE High Pressure Diffuser, 50 LPM - For 3423 and 3425 Particle Counters
Catalog No. NC3036557
Encompass
The MET ONE High Pressure Diffuser (Beckman Coulter part number 2080732-12) permits direct pressurized sampling of compressed air, nitrogen, or inert gases for Met One particle counters. It reduces sample gas pressure to atmospheric levels and supplies a flow equal to the instrument sample flow, enabling controlled sampling from pressurized sources.
  • Provides a 50 LPM sample flow for high-pressure sampling.
  • Connects via a 3/8-in. barb to the instrument.
  • Reduces pressurized sample gas to atmospheric pressure.
  • Constructed from nickel-plated aluminum and stainless steel.
  • Compact dimensions: 19.1 x 5.7 cm (7.5 x 2.25 in.).
  • Lightweight design at 0.57 kg (1.25 lb).
  • Operates at 30-150 psig (206-1034 kPa).
  • Uses a sapphire orifice for durability at the inlet.
  • Includes a stainless #4 VCR inlet connector and 1/4-in. NPT exhaust port.
  • Suitable for dry, inert, non-toxic sample gases.
Catalog No. NC3036557 Supplier Beckman Coulter Supplier No. 208073212
The MET ONE High Pressure Diffuser (Beckman Coulter part number 2080732-12) permits direct pressurized sampling of compressed air, nitrogen, or inert gases for Met One particle counters. It reduces sample gas pressure to atmospheric levels and supplies a flow equal to the instrument sample flow, enabling controlled sampling from pressurized sources.
  • Provides a 50 LPM sample flow for high-pressure sampling.
  • Connects via a 3/8-in. barb to the instrument.
  • Reduces pressurized sample gas to atmospheric pressure.
  • Constructed from nickel-plated aluminum and stainless steel.
  • Compact dimensions: 19.1 x 5.7 cm (7.5 x 2.25 in.).
  • Lightweight design at 0.57 kg (1.25 lb).
  • Operates at 30-150 psig (206-1034 kPa).
  • Uses a sapphire orifice for durability at the inlet.
  • Includes a stainless #4 VCR inlet connector and 1/4-in. NPT exhaust port.
  • Suitable for dry, inert, non-toxic sample gases.