Learn More
TA Instruments High Pressure High Temperature Capsule Seals, Multiple Layered Seals, Seal High Pressure High Temperature Capsules, One Time Use, Temperature Ambient to 550°C, Pressure 2900 Psig to 6 MPa, 20 Pack
Catalog No. 5000580851 Shop All TA Instruments Products
Encompass_Preferred
Waters TA Instruments High Pressure High Temperature Capsule Seals are multiple layered seals used to seal high-pressure high-temperature capsules. Designed for one-time use. Temperature range ambient to 550°C. Pressure range 2900 psig at up to 380°C; 6 MPa at 550°C.
Catalog No. 50-005-80851 Supplier TA Instruments Supplier No. 900794.901
Waters TA Instruments High Pressure High Temperature Capsule Seals are multiple layered seals used to seal high-pressure high-temperature capsules. Designed for one-time use. Temperature range ambient to 550°C. Pressure range 2900 psig at up to 380°C; 6 MPa at 550°C.