Learn More
TA Instruments High Pressure + High Temperature Capsule Seals, Package of 20, One-Time Use, Multi-Layered Seals, Temp Range Ambient to 550°C, Pressure Range 2900 psig (20 MPa) at 380°C, 6 MPa at 550°C, Compatible with P/N 900812.901
Catalog No. 5000580852 Shop All TA Instruments Products
Encompass_Preferred
Waters TA Instruments High Pressure + High Temperature Capsule Seals (pkg. of 20) are multi-layered seals designed for one-time use in high-pressure DSC experiments. These seals are compatible with capsule part number 900812.901 and support temperature ranges from ambient to 550 °C. Pressure capabilities include up to 2900 psig (20 MPa) at 380 °C and 6 MPa at 550 °C. They ensure secure sealing and reliable performance under demanding thermal and pressure conditions.
Catalog No. 50-005-80852 Supplier TA Instruments Supplier No. 900811.901
Waters TA Instruments High Pressure + High Temperature Capsule Seals (pkg. of 20) are multi-layered seals designed for one-time use in high-pressure DSC experiments. These seals are compatible with capsule part number 900812.901 and support temperature ranges from ambient to 550 °C. Pressure capabilities include up to 2900 psig (20 MPa) at 380 °C and 6 MPa at 550 °C. They ensure secure sealing and reliable performance under demanding thermal and pressure conditions.