Learn More
TA Instruments High Pressure High Temperature Pans, Five Sets DSC Capsules, Pans and Lids, Temperature Ambient to 550°C, Pressure 2900 Psig to 6 MPa, High Pressure High Temperature Applications, 5 Pack
Catalog No. 5000580664 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments High Pressure High Temperature Pans are five sets of DSC high-pressure high-temperature capsules with pans and lids. Temperature range ambient to 550°C. Pressure range 2900 psig at up to 380°C; 6 MPa at 550°C.
Catalog No. 50-005-80664 Supplier TA Instruments Supplier No. 900793.901
Waters TA Instruments High Pressure High Temperature Pans are five sets of DSC high-pressure high-temperature capsules with pans and lids. Temperature range ambient to 550°C. Pressure range 2900 psig at up to 380°C; 6 MPa at 550°C.