Learn More
TA Instruments High Pressure + High Temperature Pans, Package of 5 Sets, Includes Pans and Lids, Temp Range Ambient to 550°C, Pressure Range 2900 psig (20 MPa) at 380°C, 6 MPa at 550°C, Compatible with High Pressure DSC Applications
Catalog No. 5000580667 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments High Pressure + High Temperature Pans (pkg. of 5) includes five sets of DSC capsules designed for high-pressure and high-temperature applications. Each set contains a pan and lid capable of operating from ambient to 550 °C. These capsules support pressure conditions up to 2900 psig (20 MPa) at 380 °C and 6 MPa at 550 °C. Designed for use with Waters TA Instruments pressure DSC systems, they ensure safe and accurate thermal analysis under elevated conditions.
Catalog No. 50-005-80667 Supplier TA Instruments Supplier No. 900812.901
Waters TA Instruments High Pressure + High Temperature Pans (pkg. of 5) includes five sets of DSC capsules designed for high-pressure and high-temperature applications. Each set contains a pan and lid capable of operating from ambient to 550 °C. These capsules support pressure conditions up to 2900 psig (20 MPa) at 380 °C and 6 MPa at 550 °C. Designed for use with Waters TA Instruments pressure DSC systems, they ensure safe and accurate thermal analysis under elevated conditions.