Learn More
TA Instruments High Pressure + High Temperature Sample Pan Kit, Includes Plate Die, Socket Drive, Coated Seals (20), Reusable Pans (5), Tweezer, Manuals, Temp Range 380–550°C, Pressure Range 6–20 MPa, Compatible with High Pressure DSC Applications
Catalog No. 5000580767 Shop All TA Instruments Products
Encompass_Preferred
Waters TA Instruments High Pressure + High Temperature Sample Pan Kit includes all components required for high-pressure DSC experiments. The kit contains a plate die, socket drive, 20 seals coated for high-temperature and high-pressure use, 5 reusable pans, stainless steel tweezers, and manuals. Designed for experiments ranging from 380 °C to 550 °C and pressures from 6 MPa to 20 MPa, this kit supports reliable sealing and sample containment under extreme conditions. Ideal for use with Waters TA Instruments pressure DSC modules.
Catalog No. 50-005-80767 Supplier TA Instruments Supplier No. 900803.901
Waters TA Instruments High Pressure + High Temperature Sample Pan Kit includes all components required for high-pressure DSC experiments. The kit contains a plate die, socket drive, 20 seals coated for high-temperature and high-pressure use, 5 reusable pans, stainless steel tweezers, and manuals. Designed for experiments ranging from 380 °C to 550 °C and pressures from 6 MPa to 20 MPa, this kit supports reliable sealing and sample containment under extreme conditions. Ideal for use with Waters TA Instruments pressure DSC modules.