Learn More
TA Instruments Aluminum Lids (pkg. of 200), Temperature Range: -180°C to 600
Catalog No. 5000580589 Shop All Waters TA Instruments Products
Encompass_Preferred
New lightweight, quality pans for analysis of a wide variety of samples and encapsulation of materials in the crimp seal mode. These lids will work with the Tzero or Standard DSC Press.
Catalog No. 50-005-80589 Supplier TA Instruments Supplier No. 900779.901
New lightweight, quality pans for analysis of a wide variety of samples and encapsulation of materials in the crimp seal mode. These lids will work with the Tzero or Standard DSC Press.