Learn More
Grainger High Pressure Inflating Connection Bent
Catalog No. 502750785
Encompass_Preferred
The Haltec High Pressure Inflating Connection is a high pressure inflating connection for use with air compressors and air tools. It has a 1/4" NPT female inlet and a 1/4" NPT male outlet. The connection is made of brass and is rated for working pressures up to 300 PSI. The connection allows you to connect air hoses and air tools to an air compressor.
Catalog No. 50-275-0785 Supplier Grainger Supplier No. 140Y62
The Haltec High Pressure Inflating Connection is a high pressure inflating connection for use with air compressors and air tools. It has a 1/4" NPT female inlet and a 1/4" NPT male outlet. The connection is made of brass and is rated for working pressures up to 300 PSI. The connection allows you to connect air hoses and air tools to an air compressor.