Learn More
Vici Valco HIGH PRESSURE PEEK TEE,THE 1/1
SDP
Catalog No. NC2197614
Encompass_Preferred
High Pressure PEEK Tee,The 1/16'' tee comes complete with fingertight nuts, Size: 1/16", bore, 1 mm
This item is not returnable. View return policy
Catalog No. NC2197614 Supplier Vici Valco Supplier No. ZT1LFPK
High Pressure PEEK Tee,The 1/16'' tee comes complete with fingertight nuts, Size: 1/16", bore, 1 mm