Description
- The extra force makes the pump ideal for delivery of fluid to reactors in chemical applications or for working with viscous fluids.
- Robust design of the syringe holder ensures the syringe is kept level during delivery of the fluid
- features two different types of clamping mechanisms, for both smaller and larger syringes
- When the application demands a more rugged pump, this high pressure syringe pump
Specifications
Specifications
| For Use With (Application) | Neuroscience |
| Model | High pressure; Single syringe |