Learn More
Waters Corp High Pressure Seal, Flanged, 0.125 in. ID, 2 per Package
Catalog No. 502392087 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This product is a replacement high pressure seal designed for use in ACQUITY Arc and Arc HPLC Quaternary Solvent Managers.
  • Used as a component in ACQUITY UPLC QSM.
  • Included in rebuild kits.
  • Package contains 2 units.
  • Functions as a pump head and is part of a rebuild kit.
  • Compatible with HPLC and UPLC system types.
  • Recommended for annual replacement.
Catalog No. 50-239-2087 Supplier Waters Corp Supplier No. 700010663
This product is a replacement high pressure seal designed for use in ACQUITY Arc and Arc HPLC Quaternary Solvent Managers.
  • Used as a component in ACQUITY UPLC QSM.
  • Included in rebuild kits.
  • Package contains 2 units.
  • Functions as a pump head and is part of a rebuild kit.
  • Compatible with HPLC and UPLC system types.
  • Recommended for annual replacement.