Learn More
Waters Corp Waters Corporation Flowcell, 16.3 μL, 10 mm, 3000 psi
Catalog No. 50793749 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This flowcell is engineered for high-pressure applications in analytical systems. It provides essential performance characteristics for precise detection.
  • Volume of 16.3 μL
  • Pathlength of 10 mm
  • Maximum pressure of 3000 psi
  • High pressure design
  • Taper slit configuration
Catalog No. 50-793-749 Supplier Waters Corp Supplier No. WAT081321
This flowcell is engineered for high-pressure applications in analytical systems. It provides essential performance characteristics for precise detection.
  • Volume of 16.3 μL
  • Pathlength of 10 mm
  • Maximum pressure of 3000 psi
  • High pressure design
  • Taper slit configuration