Learn More
Leica Biosystems High-profile disposable blades
Catalog No. 502552455
Encompass_Preferred
When your application calls for thicker sections, look to the high-profile disposable blades Leica 818 with sharpness, longevity, and consistency.The rectangular shaped corners allow the microtomist to use the full length of the microtome blade for sectioning. The dispenser alignment pin allows direct loading into suitable blade holders.For use with Leica Biosystems microtomes classified and labeled as medical devices for histological medical diagnosis, e.g. cancer diagnosis.For in Vitro Diagnostic Use
This item is not returnable. View return policy
Catalog No. 50-255-2455 Supplier Leica Biosystems Supplier No. 14035838383
When your application calls for thicker sections, look to the high-profile disposable blades Leica 818 with sharpness, longevity, and consistency.The rectangular shaped corners allow the microtomist to use the full length of the microtome blade for sectioning. The dispenser alignment pin allows direct loading into suitable blade holders.For use with Leica Biosystems microtomes classified and labeled as medical devices for histological medical diagnosis, e.g. cancer diagnosis.For in Vitro Diagnostic Use