Learn More
Mopec™ High Profile Microtome Blade
SDP
Catalog No. 22445714 Shop All Mopec Products
Change view
Click to view available options
Profile:
High
Catalog No. Profile
22-445-714 High
1 options
Catalog No. 22-445-714 Supplier Mopec™ Supplier No. BM100HP
Add to Cart
Add to Cart
These microtome blades are complete with a composite coating on each blade.
  • Coating specifically engineered to yield high-quality sections with enhanced longevity to extend use
Specifications
Profile High
For Use With (Equipment) High Profile Microtome
Quantity 50 Blades/Pk.