Learn More
Description
- Free of detectable DNase, RNase, human DNA
- Non-Pyrogenic, non-cytotoxic
- Offer protection against contamination and evaporation during sample storage and cell cultivation
Specifications
Specifications
| Color | Clear |
| Dimensions (L x W x H) | 128 x 85 x 10 mm |
| Material | Polystyrene |
| Sterility | Sterile |
| No. of Wells | 96 |
| Condensation Rings | No |
| No. per Case | 100 |
| No. per Pack | 1 |
| Product Type | Microplate Lid |
| Adhesive | None |
| Show More |
For Research Use Only. Not for use in diagnostic procedures, unless otherwise indicated.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
