Learn More
Sigma Aldrich Fine Chemicals Biosciences High Pure PCR Product Puri
Catalog No. 502905235
Encompass
The High Pure PCR Product Purification Kit efficiently and conveniently isolates PCR products from amplification reactions and purifies nucleic acids from other modification reactions. It is also recommended for the purification of cDNA.
Catalog No. 50-290-5235 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 11732676001
The High Pure PCR Product Purification Kit efficiently and conveniently isolates PCR products from amplification reactions and purifies nucleic acids from other modification reactions. It is also recommended for the purification of cDNA.