Learn More
Sigma Aldrich Fine Chemicals Biosciences High Pure Plasmid Isolatio
Catalog No. 502904411
Encompass
Nucleic acids bind to the surface of the glass fiber fleece in the presence of a chaotropic salt (guanidine HCl). This allows the High Pure filter tube to specifically immobilize nucleic acids (both DNA and RNA) while they are freed of contaminants.Capacity The High Pure Spin filter Tubes hold up to 700 muL sample volume.Sample 0.5 - 4.0 mL cultures of E. coli (harvested at a density of 1.5 - 5.0 A 600 units/mL)
Catalog No. 50-290-4411 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 11754785001
Nucleic acids bind to the surface of the glass fiber fleece in the presence of a chaotropic salt (guanidine HCl). This allows the High Pure filter tube to specifically immobilize nucleic acids (both DNA and RNA) while they are freed of contaminants.Capacity The High Pure Spin filter Tubes hold up to 700 muL sample volume.Sample 0.5 - 4.0 mL cultures of E. coli (harvested at a density of 1.5 - 5.0 A 600 units/mL)