Learn More
Yamato Scientific America, Inc. High Purity DI System - 2-1/2" Diameter (10" Carbon, (2) 20" Mixed Bed, & 200 K DI Light). Type 2 and 3 water quality.
Catalog No. NC2334259 Shop All Yamato Scientific America Inc. Products
Encompass_Preferred
A pre-assembled cartridge system that provides deionized water using a staged filtration process. Water purification is provided using a three-stage process.
Catalog No. NC2334259 Supplier Yamato Scientific America, Inc. Supplier No. WH201P
A pre-assembled cartridge system that provides deionized water using a staged filtration process. Water purification is provided using a three-stage process.