Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ High-Purity Glucoside Detergents

Catalog No. FERA65516 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 g
Reagent Type:
Octyl-beta-Glucoside
Octylthioglucoside (OTG)
n-Nonyl-β-D-glucoside (NG)
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Reagent Type
FERA65516 5 g n-Nonyl-β-D-glucoside (NG)
PI28310 5 g Octyl-beta-Glucoside
PI28351 5 g Octylthioglucoside (OTG)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. FERA65516 Supplier Thermo Scientific™ Supplier No. A65516
Season of Savings
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Glucoside detergents are a class of non-ionic detergents commonly used in biochemical and biophysical research. These detergents are derived from glucose and possess unique properties that make them suitable for various applications.

Glucoside detergents are a class of non-ionic detergents commonly used in biochemical and biophysical research. These detergents are derived from glucose and possess unique properties that make them suitable for various applications. Glucoside detergents are known for their mildness, biocompatibility, and ability to solubilize and stabilize a wide range of biomolecules, including membrane proteins. Glucoside detergents are also compatible with many downstream applications, such as protein purification, crystallization, and structural studies.

Features of glucoside detergents:

  • Gentle on proteins, minimizing denaturation
  • Non-ionic nature—reduces electrostatic interactions
  • Low critical micelle concentration (CMC)
  • High-purity with low UV absorptivity

Overall, glucoside detergents are versatile and can be used in various experimental techniques, including protein purification, crystallization, and structural biology studies. They are compatible with a wide range of biomolecules and can be employed in both membrane and soluble protein research. With their gentle nature and versatile performance, glucoside detergents have become valuable tools in the field of biochemical research.

Octyl-beta-Glucoside

Octyl-beta-Glucoside (OG) is a low molecular weight, non-ionic detergent with an octyl chain attached to the glucose moiety that has been widely used for membrane protein solubilization. OG is generally more soluble in aqueous solutions and forms clear and stable solutions at higher concentrations. OG typically forms larger micelles compared to OTG. The larger micelles of OG can be advantageous for certain applications, such as protein purification or structural studies, as they provide better solubility and reduced interference during downstream processes.

Properties of OG

  • Alternative names: octyl-beta-glucopyranoside, octyl-beta-D-glucopyranoside
  • Molecular weight: 292.37 g
  • Micelle molecular weight: 8000 g
  • Critical micelle concentration (CMC): 23 to 25 mM (0.6716 to 0.7300%, w/v)
  • Aggregation number: 27
  • Cloud point: >100°C
  • Optically clear: low absorbance at 280 nm

Octylthio Glucoside

Octylthio Glucoside (OTG) is a low molecular weight, nonionic detergent that is effective for cell lysis and nondenaturing protein solubilization. It is resistant to beta-D-glucoside glucohydrolase degradation because the thioether group present in OTG provides protection against enzymatic hydrolysis. As a result, OTG can remain stable in the presence of beta-D-glucoside glucohydrolase enzymes, making it useful in applications where enzymatic stability is desired. However, it is important to note that the specific stability of OTG can still be influenced by factors such as enzyme concentration, reaction conditions, and the specific enzymes present.

Properties of OTG

  • Chemical name: n-octyl-β-D-thioglucopyranoside
  • Molecular weight: 44 g
  • CMC: 9 mM (0.2772%, w/v)
  • Cloud point: >100°C
  • Optically clear: low absorbance at 280 nm

n-Nonyl-β-D-glucoside

n-Nonyl-β-D-glucoside (NG) has a longer alkyl chain (nonyl) compared to other glucosides. The longer alkyl chain in n-Nonyl-β-D-glucoside can confer higher hydrophobicity and potentially stronger detergent properties. This can enhance the solubilization of hydrophobic biomolecules and improve the extraction of membrane proteins. It may also induce different interactions with lipid bilayers, influencing the stability, fluidity, and permeability of the lipid bilayer, potentially impacting the behavior of membrane proteins. In addition, NG forms larger micelles compared to glucosides with shorter alkyl chains.

Properties of NG

  • CMC: (H2O) ~ 6.5 mM (0.20%)
  • Aggregation number: ~ 133
  • Purity: ≥ 99% β+α (by HPLC analysis)
  • pH: 5-8 (1% solution in water)
TRUSTED_SUSTAINABILITY

Specifications

Color White
Product Type Detergent
CAS 69984-73-2
Molecular Formula C15H30O6
Solubility ≥20% (in water at 0°C to 5°C)
Content And Storage Store below -20°C.
Form Lyophilized
Reagent Type n-Nonyl-β-D-glucoside (NG)
Quantity 5 g
Molecular Weight (g/mol) 308.44

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.