Learn More
Perkin Elmer US LLC High Purity Quartz Baffled Cyclonic Spray Chamber with Gas Port for NexION 300/350 PC3 Systems
Catalog No. 502104910
Encompass_Preferred
The baffled cyclonic spray chamber is an all-purpose spray chamber for non-hydrofluoric acid samples. It features O-ring-free connections to the nebulizer and torch injector, secure, leak-free threaded drain port, 7 mm internal baffle, and threaded auxiliary port for gas addition. The spray chamber provides excellent sensitivity, stability and low blanks.
Catalog No. 50-210-4910 Supplier Perkin Elmer US LLC Supplier No. N8145120
The baffled cyclonic spray chamber is an all-purpose spray chamber for non-hydrofluoric acid samples. It features O-ring-free connections to the nebulizer and torch injector, secure, leak-free threaded drain port, 7 mm internal baffle, and threaded auxiliary port for gas addition. The spray chamber provides excellent sensitivity, stability and low blanks.