Learn More
Sigma Aldrich Fine Chemicals Biosciences High purity zinc NISTR SRM
Catalog No. 502907192
Encompass
This Standard Reference Material (SRM) is intended primarily for the calibration of instruments and the evaluation of chemical methods used in zinc analysis. SRM 682 is in the form of a semicircular bar segment 57 mm diamter ( 2 1/4 inch) 25.4 mm (1 inch) deep at mid diameter and 19 mm (3/4 inch) long. For more information please refer to the COA and SDS.SRM 682 cert SRM 682 SDS
Catalog No. 50-290-7192 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. NIST682
This Standard Reference Material (SRM) is intended primarily for the calibration of instruments and the evaluation of chemical methods used in zinc analysis. SRM 682 is in the form of a semicircular bar segment 57 mm diamter ( 2 1/4 inch) 25.4 mm (1 inch) deep at mid diameter and 19 mm (3/4 inch) long. For more information please refer to the COA and SDS.SRM 682 cert SRM 682 SDS