Learn More
CHEMetrics™ by AquaPhoenix High Range Nitrate Comparator Flat

Description
- C-6908 Nitrate High Range Comparator is a component of K-6901, K-6901A, and K-6901D Nitrate CHEMets Visual Test Kit and is designed to be compared against the color formed in R-6901 Nitrate CHEMets Refill.
- The comparator's range is applicable to many industrial and environmental applications.
Specifications
Specifications
| pH Range | 7 |
| Volume (Metric) | 0.5 mL |
| For Use With (Equipment) | K-6901, K-6901A or K-6901D |
| Type | Nitrate Comparator |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.