Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHEMetrics™ by AquaPhoenix High Range Phosphate, Ortho Comparator
SDP

Catalog No. 13299613
Change view
Click to view available options
Range:
1 to 10 ppm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Range
13-299-613 1 to 10 ppm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-299-613 Supplier CHEMetrics™ by AquaPhoenix Supplier No. C8510
Add to Cart
Add to Cart

High range ortho phosphate comparator designed for comparison against color formed during the reaction. Supports visual analysis of phosphate test results.

  • The C-8510 Flat Phosphate Comparator is a component of K-8510 and K-8520D phosphate test kits.
  • It is designed to be compared against the color formed in R-8510 Phosphate, ortho CHEMets™ Refill. The comparator's range is applicable for many industrial water, wastewater, and environmental water quality applications. The color standards contain blue dye mixtures that are formulated to match the colors produced by the test ampoules.

Specifications

pH Range 7
Volume (Metric) 0.5 mL
For Use With (Equipment) K-8510 or K-8520D
Range 1 to 10 ppm
Type Phosphate Comparator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.