Learn More
Electron Microscopy Sciences SEM High Resolution Au on Carbon Test Specimen, Hitachi Stub
SDP
Catalog No. 5026512
Encompass_Preferred
The gold on carbon specimen is for resolution test of SE and BSE imaging and also for chemical mapping in high-resolution systems such as an Auger scanning instrument. Each test specimen has a square grid pattern with large crystals in the center of each square, and very fine crystals at the edges of each grid somedium and high resolution gap tests are performed on the same specimen. The larger crystals facets are used to assess the gray level reproduction at high resolution. Gold particles range from approximately 5nm to 150 nm.
Mount: Hitachi Stub
Catalog No. 50-265-12 Supplier Electron Microscopy Sciences Supplier No. 7951004
The gold on carbon specimen is for resolution test of SE and BSE imaging and also for chemical mapping in high-resolution systems such as an Auger scanning instrument. Each test specimen has a square grid pattern with large crystals in the center of each square, and very fine crystals at the edges of each grid somedium and high resolution gap tests are performed on the same specimen. The larger crystals facets are used to assess the gray level reproduction at high resolution. Gold particles range from approximately 5nm to 150 nm.
Mount: Hitachi Stub