Learn More
Molecular Research Center HIGH SALT PRECIP 100ML
SDP
Catalog No. NC9549521
Encompass
HIGH SALT PRECIPITATION SOLUTION,100 ml,molecular biology grade solution of sodium chloride and sodium citrate specially designed for RNA precipitation in the TRI Reagent, procedure,Contains:1.2M NaCl and 0.8M C6H5Na3O7.
Catalog No. NC9549521 Supplier Molecular Research Center Supplier No. PS 161
HIGH SALT PRECIPITATION SOLUTION,100 ml,molecular biology grade solution of sodium chloride and sodium citrate specially designed for RNA precipitation in the TRI Reagent, procedure,Contains:1.2M NaCl and 0.8M C6H5Na3O7.