Learn More
Lakeside Manufacturing Inc High-Security Storage Cabinet, (3) Fixed Shelves
Catalog No. 502884406
Encompass_Preferred
Control access to your high-cost medications and supplies. The Lakeside LHS-120 High-Security Storage Cabinet has (3) Fixed Shelves. This unit has a heavy steel construction with a double wall door. Electronic lock with backup battery (self-contained). Different shelf configurations to fit your needs. Key lock override with 250+ possible entry codes. Made in the USA.
Catalog No. 50-288-4406 Supplier Lakeside Manufacturing Inc Supplier No. LHS120
Control access to your high-cost medications and supplies. The Lakeside LHS-120 High-Security Storage Cabinet has (3) Fixed Shelves. This unit has a heavy steel construction with a double wall door. Electronic lock with backup battery (self-contained). Different shelf configurations to fit your needs. Key lock override with 250+ possible entry codes. Made in the USA.