Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ High Select™ Depletion Spin Columns

Catalog No. PIA36368 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 Columns
24 Columns
50 mL
6 Columns
Product Type:
Midi Spin Column
Mini Spin Column
Resin
Purification Target:
HSA/Immunoglobulin
Top14 Abundant Protein
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type Purification Target
PIA36368 50 mL Resin HSA/Immunoglobulin
PIA36365 6 Columns Mini Spin Column HSA/Immunoglobulin
PIA36366 24 Columns Mini Spin Column HSA/Immunoglobulin
PIA36367 10 Columns Midi Spin Column HSA/Immunoglobulin
PIA36371 10 Columns Midi Spin Column Top14 Abundant Protein
PIA36372 50 mL Resin Top14 Abundant Protein
PIA36369 6 Columns Mini Spin Column Top14 Abundant Protein
PIA36370 24 Columns Mini Spin Column Top14 Abundant Protein
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. PIA36368 Supplier Thermo Scientific™ Supplier No. A36368
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Decrease albumin, immunoglobulins, and other high-abundance proteins from human serum or plasma in preparation for mass spectrometry analysis, or 1D and 2D gel electrophoresis using High Select depletion spin columns or resin.

Analysis of human fluids is often complicated by the presence of high concentrations of serum proteins, including albumin and IgG that can make up more than 70% of total serum protein. Removal of albumin, immunoglobulins, and other abundant proteins is essential for the study of low-abundance proteins. Thermo Scientific High Select HSA/Immunoglobulin and Top 14 Abundant Protein depletion spin columns and resins enable depletion of the most abundant proteins in human serum and plasma for improved detection of low-abundance proteins using mass spectrometry analysis or 1D or 2D gel electrophoresis.

Flexible—resin provided in mini and midi column formats as well as bulk for custom filling options
Optimized—removes >95% of IgG and >95% of albumin (HSA/Immunoglobulin); removes >95% of IgG and >95% of albumin, plus 12 other abundant proteins (Top 14)
Fast—process samples in ∼10 minutes
Economical—cost-effective spin columns are priced for single use
Reduced carry-over—one-time use prevents protein carryover and experimental variability

High Select HSA/Immunoglobulin Depletion Resin uses highly specific, immobilized anti-HSA and anti-immuoglobulin antibodies (IgG, IgA, IgM, IgD, and IgE) to remove human serum albumin (HSA) and all major subclasses of immunoglobulins from serum and plasma. High Select Top14 Abundant Protein Depletion Resin uses highly specific immobilized antibodies to remove human serum albumin (HSA), albumin, IgG, IgA, IgM, IgD, IgE, kappa and lambda light chains, alpha-1-acidglycoprotein, alpha-1-antitrypsin, alpha-2-macroglobulin, apolipoprotein A1, fibrinogen, haptoglobin, and transferrin from serum, plasma, or spinal fluids. The resins are provided in an economical and convenient spin column format specifically designed for one-step processing and single use or bulk format for custom filling.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Mass Spectrometer
Product Type Resin
Product Line High Select
Content And Storage Store at 2–8°C.
Label or Dye Unlabeled
Shipping Condition Wet Ice
Starting Material Plasma, Serum
Final Product Type Protein
Workflow Step Protein Depletion
Quantity 50 mL
Purification Target HSA/Immunoglobulin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.