Learn More
Thermo Scientific Thermo Scientific High Sensitivity D2 Lamp VF-D4 Detector
Catalog No. NC1576292 Shop All Thermo Scientific Products
Encompass
High sensitivity deuterium lamp emits 190-400nm UV light with optimal intensity as light source for spectrophotometry and HPLC systems.
Catalog No. NC1576292 Supplier Thermo Scientific Supplier No. 6077.1111
High sensitivity deuterium lamp emits 190-400nm UV light with optimal intensity as light source for spectrophotometry and HPLC systems.